siRNA Details
Gene Symbol: FCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK314867.1 | - | 2 | 245 | ||
BC020635.1 | - | 2 | 227 | 4717731 | ||
BG482630.1 | - | 2 | 296 | |||
CR749452.1 | - | 2 | 213 | |||
D83920.1 | - | 2 | 201 | |||
DA813504.1 | - | 2 | 339 |
