siRNA Details
Gene Symbol: GPALPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316951.1 | NP_001303880.1 | 5 | 593 | ||
NM_001316952.1 | NP_001303881.1 | 5 | 714 | |||
NM_018559.3 | NP_061029.2 | 5 | 593 | |||
XM_005266444.3 | XP_005266501.1 | 5 | 585 | |||
XM_011535144.2 | XP_011533446.1 | 5 | 652 | |||
GenBank mRNA | AB051491.1 | - | 5 | 503 | ||
AF145122.1 | - | 5 | 527 | |||
AF245045.1 | - | 4 | 379 | |||
AK292493.1 | - | 5 | 566 | |||
AK304669.1 | - | 5 | 603 | |||
BC017776.1 | - | 5 | 578 | |||
BC032333.1 | - | 5 | 640 | 5169953 | ||
BC039586.1 | - | 5 | 571 | 5722236 |
