siRNA Details
Gene Symbol: LANCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018697.3 | NP_061167.1 | 4, 5 | 1264 | ||
GenBank mRNA | AB035966.1 | - | 4, 5 | 741 | ||
AF353942.1 | - | 4, 5 | 741 | |||
AJ278245.1 | - | 4, 5 | 872 | |||
AK095857.1 | - | 4, 5 | 1264 | |||
AK225958.1 | - | 4, 5 | 717 | |||
AK301747.1 | - | 4, 5 | 670 | |||
AK313326.1 | - | 4, 5 | 1009 | |||
AL713665.2 | - | 4, 5 | 656 | |||
BC004887.2 | - | 1, 2 | 92 | |||
BC070049.1 | - | 4, 5 | 960 | 30343188 |
