siRNA Details
Gene Symbol: GFOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242628.1 | NP_001229557.1 | 2 | 1242 | ||
NM_001242630.1 | NP_001229559.1 | 2 | 1297 | |||
NM_018988.3 | NP_061861.1 | 2 | 2069 | |||
XM_011514699.2 | XP_011513001.1 | 2 | 1258 | |||
GenBank mRNA | AK000337.1 | - | 2 | 1987 | ||
AK027010.1 | - | 2 | 1297 | |||
AK055131.1 | - | 2 | 1211 | |||
BC119005.1 | - | 2 | 1409 | 40085551 | ||
BC124557.1 | - | 2 | 1409 | 40085552 | ||
BC148295.2 | - | 2 | 1416 | 40122213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
