siRNA Details
Gene Symbol: AKR1B15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080538.2 | NP_001074007.2 | 12 | 1343 | ||
XM_011516241.2 | XP_011514543.2 | 10 | 1745 | |||
XM_017012224.1 | XP_016867713.1 | 10 | 1096 |
Gene Symbol: AKR1B10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020299.4 | NP_064695.3 | 10 | 1280 | ||
XM_011516416.1 | XP_011514718.1 | 9 | 1155 | |||
GenBank mRNA | AF044961.1 | - | 4 | 268 | ||
AF052577.1 | - | 10 | 999 | |||
AF524864.1 | - | 10 | 1280 | |||
BC008837.2 | - | 10 | 1225 | 3639480 | ||
U37100.1 | - | 10 | 1029 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
