siRNA Details
Gene Symbol: ZNF286A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130842.1 | NP_001124314.1 | 6 | 1404 | ||
NM_001288642.1 | NP_001275571.1 | 6 | 1331 | |||
NM_001288643.1 | NP_001275572.1 | 5 | 1172 | |||
NM_001288644.1 | NP_001275573.1 | 4 | 1057 | |||
NM_001288645.1 | NP_001275574.1 | 6 | 1550 | |||
NM_001288646.1 | NP_001275575.1 | 6 | 1463 | |||
NM_001288647.1 | NP_001275576.1 | 5 | 1348 | |||
NM_001288648.1 | NP_001275577.1 | 5 | 1231 | |||
NM_001288649.1 | NP_001275578.1 | 4 | 1052 | |||
NM_020652.2 | NP_065703.1 | 6 | 1491 | |||
GenBank mRNA | AF217226.1 | - | 5 | 1323 | ||
AK092259.1 | - | 6 | 1388 | |||
AK315062.1 | - | 6 | 1249 | |||
BC015730.1 | - | 6 | 1388 | |||
BC025260.1 | - | 6 | 1388 | |||
BC128259.1 | - | 5 | 1218 | 40116891 | ||
BC128411.1 | - | 5 | 1218 | 40116881 |
Gene Symbol: ZNF286B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145045.1 | NP_001138517.1 | 5 | 1262 |
