siRNA Details
Gene Symbol: ZNF286A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130842.1 | NP_001124314.1 | 6 | 2066 | ||
NM_001288642.1 | NP_001275571.1 | 6 | 1993 | |||
NM_001288643.1 | NP_001275572.1 | 5 | 1834 | |||
NM_001288644.1 | NP_001275573.1 | 4 | 1719 | |||
NM_001288645.1 | NP_001275574.1 | 6 | 2212 | |||
NM_001288646.1 | NP_001275575.1 | 6 | 2125 | |||
NM_001288647.1 | NP_001275576.1 | 5 | 2010 | |||
NM_001288648.1 | NP_001275577.1 | 5 | 1893 | |||
NM_001288649.1 | NP_001275578.1 | 4 | 1714 | |||
NM_020652.2 | NP_065703.1 | 6 | 2153 | |||
GenBank mRNA | AF217226.1 | - | 5 | 1985 | ||
AK092259.1 | - | 6 | 2050 | |||
BC015730.1 | - | 6 | 2050 | |||
BC025260.1 | - | 6 | 2050 |
