siRNA Details
Gene Symbol: PHF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020889.2 | NP_065940.1 | 9 | 2726 | ||
GenBank mRNA | AY030283.1 | - | 9 | 2728 | ||
BC121043.1 | - | 9 | 2228 | 40117907 | ||
BC121044.1 | - | 9 | 2228 | 40117911 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
