siRNA Details
Gene Symbol: CD209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144893.1 | NP_001138365.1 | 5 | 3688 | ||
NM_001144894.1 | NP_001138366.1 | 5 | 3964 | |||
NM_001144895.1 | NP_001138367.1 | 7 | 3820 | |||
NM_001144896.1 | NP_001138368.1 | 6 | 4024 | |||
NM_001144897.1 | NP_001138369.1 | 7 | 4078 | |||
NM_001144899.1 | NP_001138371.1 | 7 | 3613 | |||
NM_021155.3 | NP_066978.1 | 7 | 4096 | |||
NR_026692.1 | 6 | 4196 | ||||
XM_005272472.4 | XP_005272529.1 | 7 | 3713 | |||
GenBank mRNA | AF290886.1 | - | 7 | 4037 | ||
AK057212.1 | - | 2 | 2100 | |||
AK293089.1 | - | 7 | 4051 |
