siRNA Details
Gene Symbol: ZNF350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021632.3 | NP_067645.3 | 5 | 1928 | ||
XM_011527196.1 | XP_011525498.1 | 5 | 1972 | |||
XM_017027094.1 | XP_016882583.1 | 6 | 2084 | |||
XM_017027095.1 | XP_016882584.1 | 6 | 2245 | |||
XM_017027096.1 | XP_016882585.1 | 6 | 2105 | |||
XM_017027097.1 | XP_016882586.1 | 6 | 2118 | |||
XM_017027098.1 | XP_016882587.1 | 6 | 2205 | |||
XM_017027099.1 | XP_016882588.1 | 7 | 2228 | |||
XM_017027100.1 | XP_016882589.1 | 5 | 2087 | |||
XM_017027101.1 | XP_016882590.1 | 6 | 2071 | |||
XM_017027102.1 | XP_016882591.1 | 6 | 2117 | |||
XM_017027103.1 | XP_016882592.1 | 6 | 2205 | |||
XM_017027104.1 | XP_016882593.1 | 6 | 2209 | |||
GenBank mRNA | AF295096.1 | - | 4 | 1883 | ||
AF309561.1 | - | 5 | 1917 | |||
AK023467.1 | - | 5 | 1928 | |||
BC009921.2 | - | 4 | 1868 | 3506774 |
