siRNA Details
Gene Symbol: GRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002087.3 | NP_002078.1 | 12 | 1834 | ||
XM_005257253.1 | XP_005257310.1 | 12 | 1879 | |||
GenBank mRNA | AF055008.1 | - | 12 | 1640 | ||
AK000607.1 | - | 12 | 1618 | |||
AK023348.1 | - | 9 | 1122 | |||
AK222522.1 | - | 12 | 1618 | |||
AK296090.1 | - | 12 | 1572 | |||
AK303830.1 | - | 9 | 1144 | |||
AY124489.1 | - | 11 | 1596 | |||
BC010577.2 | - | 11 | 1588 | 3457813 | ||
M75161.1 | - | 11 | 1590 | |||
X62320.1 | - | 12 | 1618 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
