siRNA Details
Gene Symbol: CRLF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012288.2 | NP_001012288.2 | 7 | 1402 | ||
NM_022148.3 | NP_071431.2 | 8 | 1505 | |||
NR_110830.1 | 8 | 1684 | ||||
XM_011546181.2 | XP_011544483.1 | 8 | 1540 | |||
GenBank mRNA | AF142570.1 | - | 8 | 1432 | ||
AK026800.1 | - | 8 | 1686 | |||
AK225583.1 | - | 8 | 1686 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
