siRNA Details
Gene Symbol: MTRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002454.2 | NP_002445.2 | 15 | 2227 | ||
NM_024010.2 | NP_076915.2 | 15 | 2201 | |||
NR_134480.1 | 15 | 2213 | ||||
NR_134481.1 | 14 | 2138 | ||||
NR_134482.1 | 14 | 2073 | ||||
GenBank mRNA | AF025794.1 | - | 15 | 2169 | ||
AF121214.1 | - | 15 | 2201 | |||
BC054816.1 | - | 15 | 2142 | 5205285 | ||
BC109216.1 | - | 15 | 2201 | 40007734 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
