siRNA Details
Gene Symbol: TSSK1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032028.3 | NP_114417.1 | 1 | 485 | ||
GenBank mRNA | AF348076.1 | - | 1 | 377 | ||
AK122904.1 | - | 1 | 443 | |||
AK313332.1 | - | 1 | 438 | |||
AY028964.1 | - | 1 | 383 | |||
BC022515.1 | - | 1 | 423 | 4816988 | ||
DB025324.1 | - | 1 | 485 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
