siRNA Details
Gene Symbol: TSSK1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032028.3 | NP_114417.1 | 1 | 2029 | ||
GenBank mRNA | AK122904.1 | - | 1 | 1987 | ||
BC022515.1 | - | 1 | 1967 | 4816988 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
