siRNA Details
Gene Symbol: SLC2A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024938.3 | NP_001020109.1 | 12 | 1942 | ||
NM_001024939.3 | NP_001020110.1 | 12 | 1830 | |||
NM_001282864.1 | NP_001269793.1 | 11 | 2184 | |||
NM_030807.4 | NP_110434.3 | 13 | 2053 | |||
NR_104247.1 | 11 | 1789 | ||||
GenBank mRNA | AB209301.1 | - | 8 | 2415 | ||
AF443201.1 | - | 13 | 1884 | |||
AK075467.1 | - | 12 | 1718 | |||
AK092466.1 | - | 2 | 1854 | |||
BC018967.2 | - | 1 | 498 | |||
BC094735.1 | - | 12 | 1759 | 5304025 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
