siRNA Details
Gene Symbol: SLC37A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032295.3 | NP_115671.1 | 10, 11 | 1228 | ||
XM_017012713.1 | XP_016868202.1 | 12, 13 | 1720 | |||
XM_017012716.1 | XP_016868205.1 | 10, 11 | 1484 | |||
GenBank mRNA | AL136583.1 | - | 10, 11 | 1135 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
