siRNA Details
Gene Symbol: ZNF397 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032347.2 | NP_115723.1 | 6 | 1199 | ||
XM_006722558.2 | XP_006722621.1 | 5 | 10132 | |||
XM_011526229.2 | XP_011524531.1 | 6 | 10221 | |||
XM_011526230.2 | XP_011524532.1 | 5 | 10423 | |||
XM_011526231.2 | XP_011524533.1 | 5 | 10020 | |||
XM_017026042.1 | XP_016881531.1 | 5 | 10075 | |||
GenBank mRNA | AK027384.1 | - | 5 | 1084 | ||
BC006172.1 | - | 6 | 1174 | 4053312 |
