siRNA Details
Gene Symbol: ZNF333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300912.1 | NP_001287841.1 | 7 | 592 | ||
NM_032433.2 | NP_115809.1 | 7 | 574 | |||
XM_005260098.3 | XP_005260155.1 | 8 | 887 | |||
XM_006722924.2 | XP_006722987.1 | 7 | 592 | |||
XM_011528362.2 | XP_011526664.1 | 7 | 615 | |||
XM_011528363.2 | XP_011526665.1 | 9 | 1715 | |||
XM_011528365.2 | XP_011526667.1 | 9 | 1056 | |||
XM_011528366.2 | XP_011526668.1 | 8 | 884 | |||
XM_011528367.2 | XP_011526669.1 | 8 | 897 | |||
XM_011528368.2 | XP_011526670.1 | 8 | 804 | |||
XM_011528369.2 | XP_011526671.1 | 8 | 817 | |||
XM_017027367.1 | XP_016882856.1 | 7 | 591 | |||
XM_017027368.1 | XP_016882857.1 | 6 | 459 | |||
XM_017027369.1 | XP_016882858.1 | 8 | 2212 | |||
XM_017027370.1 | XP_016882859.1 | 8 | 2225 | |||
XM_017027371.1 | XP_016882860.1 | 8 | 837 | |||
GenBank mRNA | AF372702.1 | - | 7 | 531 | ||
AK094028.1 | - | 8 | 817 | |||
AK307147.1 | - | 8 | 898 | |||
AK309208.1 | - | 7 | 526 | |||
AL832544.1 | - | 1 | 66 | |||
BC048107.1 | - | 7 | 563 | |||
BC064571.1 | - | 7 | 1169 | |||
BX647475.1 | - | 7 | 535 |
