siRNA Details
Gene Symbol: ZNF333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300912.1 | NP_001287841.1 | 11 | 1006 | ||
NM_032433.2 | NP_115809.1 | 11 | 988 | |||
XM_005260098.3 | XP_005260155.1 | 12 | 1301 | |||
XM_006722924.2 | XP_006722987.1 | 11 | 1006 | |||
XM_011528362.2 | XP_011526664.1 | 11 | 1029 | |||
XM_011528363.2 | XP_011526665.1 | 13 | 2129 | |||
XM_011528365.2 | XP_011526667.1 | 13 | 1470 | |||
XM_011528366.2 | XP_011526668.1 | 12 | 1298 | |||
XM_011528367.2 | XP_011526669.1 | 12 | 1311 | |||
XM_011528368.2 | XP_011526670.1 | 12 | 1218 | |||
XM_011528369.2 | XP_011526671.1 | 12 | 1231 | |||
XM_011528371.2 | XP_011526673.1 | 5 | 1395 | |||
XM_011528372.2 | XP_011526674.1 | 4 | 324 | |||
XM_017027367.1 | XP_016882856.1 | 11 | 1005 | |||
XM_017027368.1 | XP_016882857.1 | 10 | 873 | |||
XM_017027369.1 | XP_016882858.1 | 12 | 2626 | |||
XM_017027370.1 | XP_016882859.1 | 12 | 2639 | |||
XM_017027371.1 | XP_016882860.1 | 12 | 1251 | |||
GenBank mRNA | AB058709.1 | - | 4 | 302 | ||
AF372702.1 | - | 11 | 945 | |||
AK094028.1 | - | 12 | 1231 | |||
AK122817.1 | - | 4 | 869 | |||
AK309208.1 | - | 10 | 1507 | |||
AL832544.1 | - | 5 | 480 | |||
BC048107.1 | - | 11 | 977 | |||
BX647475.1 | - | 11 | 949 |
