siRNA Details
Gene Symbol: PPIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032472.3 | NP_115861.1 | 7 | 1039 | ||
NM_130906.2 | NP_570981.1 | 7 | 1027 | |||
XM_005246651.3 | XP_005246708.1 | 7 | 958 | |||
XM_005246652.4 | XP_005246709.1 | 7 | 946 | |||
XM_011511358.2 | XP_011509660.1 | 7 | 1132 | |||
XM_017004352.1 | XP_016859841.1 | 7 | 1149 | |||
XM_017004353.1 | XP_016859842.1 | 7 | 888 | |||
XM_017004354.1 | XP_016859843.1 | 7 | 1322 | |||
XM_017004355.1 | XP_016859844.1 | 7 | 1346 | |||
XM_017004356.1 | XP_016859845.1 | 7 | 1310 | |||
XM_017004357.1 | XP_016859846.1 | 8 | 1229 | |||
XM_017004358.1 | XP_016859847.1 | 8 | 1052 | |||
XM_017004359.1 | XP_016859848.1 | 8 | 1236 | |||
XM_017004360.1 | XP_016859849.1 | 6 | 1049 | |||
GenBank mRNA | AF146799.1 | - | 7 | 896 | ||
AF251049.1 | - | 7 | 1039 | |||
AF271652.1 | - | 7 | 962 | |||
AF414127.1 | - | 6 | 963 | |||
AY257468.1 | - | 6 | 1083 | |||
BC007693.2 | - | 7 | 1468 | 3956681 | ||
BC031679.1 | - | 1 | 64 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
