siRNA Details
Gene Symbol: FOXP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244808.1 | NP_001231737.1 | 21 | 3347 | ||
NM_001244810.1 | NP_001231739.1 | 21 | 3295 | |||
NM_001244812.1 | NP_001231741.1 | 16 | 2647 | |||
NM_001244813.1 | NP_001231742.1 | 15 | 2539 | |||
NM_001244814.1 | NP_001231743.1 | 17 | 3044 | |||
NM_001244815.1 | NP_001231744.1 | 16 | 2985 | |||
NM_001244816.1 | NP_001231745.1 | 20 | 3152 | |||
NM_032682.5 | NP_116071.2 | 21 | 3247 | |||
XM_006713102.2 | XP_006713165.1 | 20 | 3208 | |||
XM_011533584.2 | XP_011531886.1 | 21 | 3318 | |||
XM_011533585.2 | XP_011531887.1 | 20 | 3170 | |||
XM_017006165.1 | XP_016861654.1 | 20 | 3089 | |||
GenBank mRNA | AF250920.1 | - | 20 | 3141 | ||
AK026898.1 | - | 1 | 47 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
