siRNA Details
Gene Symbol: IDI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033261.2 | NP_150286.1 | 5 | 819 | ||
GenBank mRNA | AK056950.1 | - | 5 | 819 | ||
BC017778.1 | - | 5 | 818 | 4291149 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
