siRNA Details
Gene Symbol: TRIM41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033549.4 | NP_291027.3 | 6 | 3132 | ||
XM_006714929.2 | XP_006714992.1 | 7 | 2554 | |||
XM_006714930.2 | XP_006714993.1 | 7 | 2548 | |||
GenBank mRNA | AB100366.1 | - | 6 | 2394 | ||
AF258579.1 | - | 2 | 1488 | |||
AK074052.1 | - | 2 | 5183 | |||
AL832960.1 | - | 5 | 2209 | |||
AX775789.1 | - | 6 | 1871 | |||
AX775791.1 | - | 6 | 1871 | |||
AX775797.1 | - | 6 | 2163 | |||
AX775799.1 | - | 6 | 3105 | |||
AX775801.1 | - | 5 | 3305 | |||
AX775803.1 | - | 5 | 2363 | |||
BC009762.2 | - | 2 | 1166 | |||
BC018765.1 | - | 6 | 2163 | |||
BC090953.1 | - | 6 | 2673 | 6017628 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
