siRNA Details
Gene Symbol: FCRL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052939.3 | NP_443171.2 | 15 | 2808 | ||
XM_006711145.1 | XP_006711208.1 | 15 | 2738 | |||
GenBank mRNA | AF459027.1 | - | 15 | 2808 | ||
AK125622.1 | - | 4 | 641 | |||
AL833391.1 | - | 9 | 3358 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
