siRNA Details
Gene Symbol: TRIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003818.2 | NP_001003818.1 | 2 | 486 | ||
NM_058166.4 | NP_477514.1 | 2 | 287 | |||
GenBank mRNA | AF220030.1 | - | 2 | 275 | ||
AK027664.1 | - | 2 | 279 | |||
AK290172.1 | - | 2 | 486 | |||
BC047564.1 | - | 2 | 465 | 4825118 | ||
CR749260.1 | - | 2 | 260 | |||
DB233046.1 | - | 2 | 287 |
Gene Symbol: TRIM6-TRIM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003819.3 | NP_001003819.1 | 2 | 486 | ||
GenBank mRNA | AB039903.1 | - | 2 | 394 | ||
BC136871.1 | - | 2 | 405 | 9020862 | ||
BC140722.1 | - | 2 | 405 | 9021590 | ||
BC144424.1 | - | 2 | 405 | |||
BG721109.1 | - | 2 | 470 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
