siRNA Details
Gene Symbol: TRIM6-TRIM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003819.3 | NP_001003819.1 | 3 | 823 | ||
GenBank mRNA | AB039903.1 | - | 3 | 731 | ||
BC136871.1 | - | 3 | 742 | 9020862 | ||
BC140722.1 | - | 3 | 742 | 9021590 | ||
BC144424.1 | - | 3 | 742 |
Gene Symbol: TRIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003818.2 | NP_001003818.1 | 3 | 823 | ||
NM_001198644.1 | NP_001185573.1 | 2 | 333 | |||
NM_058166.4 | NP_477514.1 | 3 | 624 | |||
GenBank mRNA | AF220030.1 | - | 3 | 612 | ||
AK027664.1 | - | 3 | 616 | |||
AK290172.1 | - | 3 | 823 | |||
AK298301.1 | - | 2 | 162 | |||
AK316178.1 | - | 3 | 290 | |||
AK316289.1 | - | 3 | 250 | |||
BC047564.1 | - | 3 | 802 | 4825118 | ||
BC065575.1 | - | 3 | 349 | 4609084 | ||
CR749260.1 | - | 4 | 519 | |||
DC354457.1 | - | 3 | 290 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
