siRNA Details
Gene Symbol: CD200R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138806.3 | NP_620161.1 | 4 | 598 | ||
NM_138939.2 | NP_620385.1 | 4 | 598 | |||
NM_138940.2 | NP_620386.1 | 3 | 529 | |||
NM_170780.2 | NP_740750.1 | 3 | 529 | |||
GenBank mRNA | AF283760.1 | - | 3 | 304 | ||
AF495380.1 | - | 4 | 420 | |||
AF497548.1 | - | 3 | 351 | |||
AF497549.1 | - | 4 | 420 | |||
AF497550.1 | - | 3 | 351 | |||
AK126349.1 | - | 3 | 529 | |||
AK293071.1 | - | 4 | 543 | |||
AK304426.1 | - | 2 | 400 | |||
AY284975.1 | - | 4 | 587 | |||
AY358910.1 | - | 4 | 623 | |||
BC069661.1 | - | 4 | 396 | 7262527 | ||
BC069721.1 | - | 4 | 396 | 7262539 | ||
BC069743.1 | - | 4 | 396 | 7262551 | ||
BC093890.1 | - | 3 | 375 | 7939735 | ||
BC143393.1 | - | 4 | 444 | 9051899 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
