siRNA Details
Gene Symbol: PPIL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139126.3 | NP_624311.1 | 8 | 867 | ||
GenBank mRNA | AF357880.1 | - | 8 | 800 | ||
AK092069.1 | - | 8 | 800 | |||
AK315388.1 | - | 8 | 867 | |||
BC020986.1 | - | 8 | 803 | 3852013 | ||
BC064134.1 | - | 8 | 785 | |||
BC107684.1 | - | 8 | 801 | |||
BX537536.1 | - | 8 | 802 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
