siRNA Details
Gene Symbol: ZNF480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297624.1 | NP_001284553.1 | 4 | 2496 | ||
NM_001297625.1 | NP_001284554.1 | 4 | 2498 | |||
NM_144684.3 | NP_653285.2 | 5 | 2625 | |||
XM_011526465.2 | XP_011524767.1 | 5 | 3027 | |||
GenBank mRNA | AL832560.1 | - | 4 | 2475 | ||
AY512662.1 | - | 5 | 2594 | |||
BC018783.1 | - | 5 | 2594 | 4873910 |
