siRNA Details
Gene Symbol: ZNF582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320371.1 | NP_001307300.1 | 5 | 2107 | ||
NM_144690.2 | NP_653291.1 | 5 | 1885 | |||
GenBank mRNA | AK055489.1 | - | 5 | 1804 | ||
BC101008.1 | - | 4 | 1726 | 40006516 | ||
BC101009.1 | - | 4 | 1726 | 40006517 | ||
BC101010.1 | - | 4 | 1726 | 40006519 | ||
BC101011.1 | - | 4 | 1727 | 40006520 |
