siRNA Details
Gene Symbol: FREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177704.1 | NP_001171175.1 | 4 | 796 | ||
NM_144966.5 | NP_659403.4 | 28 | 5915 | |||
XM_005251382.3 | XP_005251439.1 | 27 | 6163 | |||
XM_005251384.4 | XP_005251441.1 | 6 | 1209 | |||
XM_006716729.3 | XP_006716792.1 | 7 | 1454 | |||
XM_011517758.2 | XP_011516060.1 | 4 | 845 | |||
XM_017014316.1 | XP_016869805.1 | 27 | 6190 | |||
XM_017014317.1 | XP_016869806.1 | 28 | 5733 | |||
XM_017014318.1 | XP_016869807.1 | 28 | 6488 | |||
XM_017014319.1 | XP_016869808.1 | 29 | 6392 | |||
XM_017014320.1 | XP_016869809.1 | 28 | 6319 | |||
XM_017014321.1 | XP_016869810.1 | 27 | 5982 | |||
XM_017014322.1 | XP_016869811.1 | 27 | 5776 | |||
XM_017014323.1 | XP_016869812.1 | 27 | 5449 | |||
XM_017014324.1 | XP_016869813.1 | 27 | 6190 | |||
XM_017014325.1 | XP_016869814.1 | 27 | 6190 | |||
XM_017014326.1 | XP_016869815.1 | 26 | 5006 | |||
XM_017014327.1 | XP_016869816.1 | 23 | 4375 | |||
XM_017014328.1 | XP_016869817.1 | 27 | 6190 | |||
XM_017014329.1 | XP_016869818.1 | 27 | 6190 | |||
GenBank mRNA | AB160987.1 | - | 26 | 5099 | ||
AK058190.1 | - | 3 | 339 | |||
BC031064.1 | - | 4 | 796 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
