siRNA Details
Gene Symbol: ZFP62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172638.1 | NP_001166109.1 | 2 | 1863 | ||
NM_152283.4 | NP_689496.4 | 3 | 1936 | |||
XM_017009710.1 | XP_016865199.1 | 3 | 1966 | |||
XM_017009711.1 | XP_016865200.1 | 3 | 2014 | |||
XM_017009712.1 | XP_016865201.1 | 4 | 2039 | |||
XM_017009713.1 | XP_016865202.1 | 4 | 2087 | |||
XM_017009714.1 | XP_016865203.1 | 3 | 2020 | |||
XM_017009715.1 | XP_016865204.1 | 2 | 1973 | |||
XM_017009716.1 | XP_016865205.1 | 2 | 2265 | |||
XM_017009717.1 | XP_016865206.1 | 2 | 1936 | |||
XM_017009718.1 | XP_016865207.1 | 2 | 2265 | |||
XM_017009719.1 | XP_016865208.1 | 2 | 2265 | |||
XM_017009720.1 | XP_016865209.1 | 2 | 1904 | |||
XM_017009721.1 | XP_016865210.1 | 3 | 1946 | |||
GenBank mRNA | AK002206.1 | - | 1 | 419 | ||
AK091550.1 | - | 2 | 693 | |||
AK295714.1 | - | 3 | 1926 | |||
AK303450.1 | - | 2 | 1854 | |||
AL832408.1 | - | 1 | 2023 | |||
BC140761.1 | - | 1 | 1242 | 9021629 | ||
BC171780.1 | - | 1 | 1242 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
