siRNA Details
Gene Symbol: LRR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203467.1 | NP_982292.1 | 2, 3 | 614 | ||
NR_037792.1 | 5, 6 | 1556 | ||||
GenBank mRNA | BC030142.2 | - | 2, 3 | 550 | ||
BC093697.1 | - | 2, 3 | 316 | |||
BC107888.1 | - | 2, 3 | 363 | |||
BC112241.1 | - | 2, 3 | 316 | |||
BX648029.1 | - | 5, 6 | 1479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
