siRNA Details
Gene Symbol: HEPACAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152722.4 | NP_689935.2 | 7 | 2293 | ||
XM_005271449.2 | XP_005271506.1 | 7 | 2290 | |||
XM_017017361.1 | XP_016872850.1 | 7 | 2449 | |||
GenBank mRNA | AL834419.1 | - | 4 | 1138 | ||
AY047587.1 | - | 7 | 1934 | |||
BC037810.1 | - | 1 | 93 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
