siRNA Details
Gene Symbol: ZNF555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172775.1 | NP_001166246.1 | 4 | 680 | ||
NM_152791.4 | NP_690004.4 | 4 | 683 | |||
XM_011527716.2 | XP_011526018.1 | 4 | 683 | |||
XM_017026375.1 | XP_016881864.1 | 4 | 683 | |||
GenBank mRNA | AK093163.1 | - | 4 | 668 | ||
AK292954.1 | - | 4 | 628 | |||
AL832140.1 | - | 4 | 644 | |||
BC022022.2 | - | 4 | 634 | 4822197 | ||
BX647423.1 | - | 4 | 641 |
