siRNA Details
Gene Symbol: PAOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152911.3 | NP_690875.1 | 4 | 1012 | ||
NM_207128.2 | NP_997011.1 | 4 | 1012 | |||
NR_109764.1 | 4 | 613 | ||||
GenBank mRNA | AF226657.1 | - | 3 | 752 | ||
AF312698.1 | - | 4 | 1058 | |||
AK128001.1 | - | 4 | 896 | |||
AL834535.1 | - | 2 | 257 | |||
AY358418.1 | - | 4 | 952 | |||
AY541516.1 | - | 4 | 988 | |||
AY541520.1 | - | 1376 | ||||
AY541524.1 | - | 5 | 852 | |||
BC032778.1 | - | 4 | 1023 | 4932036 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
