siRNA Details
Gene Symbol: ZNF398 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020781.3 | NP_065832.1 | 7 | 2569 | ||
NM_170686.2 | NP_733787.1 | 6 | 2506 | |||
XM_011516439.2 | XP_011514741.1 | 7 | 2387 | |||
XM_011516440.2 | XP_011514742.1 | 6 | 2359 | |||
XM_011516441.2 | XP_011514743.1 | 6 | 2342 | |||
GenBank mRNA | AB037760.1 | - | 2 | 1527 | ||
AK290499.1 | - | 6 | 2474 | |||
AK304369.1 | - | 3 | 1661 | |||
AK304603.1 | - | 4 | 1800 | |||
AY049743.1 | - | 7 | 2569 | |||
AY049744.1 | - | 6 | 2248 | |||
BC043295.2 | - | 6 | 2494 | 5298104 |
