siRNA Details
Gene Symbol: SLCO6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289002.1 | NP_001275931.1 | 5, 6 | 1201 | ||
NM_001289004.1 | NP_001275933.1 | 4, 5 | 1015 | |||
NM_173488.4 | NP_775759.3 | 5, 6 | 1201 | |||
XM_005271874.3 | XP_005271931.1 | 5, 6 | 4398 | |||
XM_011543147.2 | XP_011541449.1 | 5, 6 | 4293 | |||
XM_011543148.2 | XP_011541450.1 | 5, 6 | 4161 | |||
XM_011543150.1 | XP_011541452.1 | 4, 5 | 411 | |||
XM_011543153.2 | XP_011541455.1 | 3, 4 | 291 | |||
XM_011543154.2 | XP_011541456.1 | 5, 6 | 4398 | |||
GenBank mRNA | AF505657.1 | - | 5, 6 | 1172 | ||
AY273897.1 | - | 5, 6 | 1164 | |||
BC034976.2 | - | 4, 5 | 1017 | 4820512 |
