siRNA Details
Gene Symbol: ZNF584 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318002.1 | NP_001304931.1 | 5 | 2266 | ||
NM_173548.2 | NP_775819.1 | 4 | 2168 | |||
NR_134461.1 | 5 | 2251 | ||||
XM_005258628.4 | XP_005258685.1 | 4 | 2647 | |||
XM_005258630.4 | XP_005258687.1 | 5 | 2284 | |||
GenBank mRNA | AK097218.1 | - | 1 | 2595 | ||
BC017651.1 | - | 5 | 2173 | |||
BC033149.1 | - | 4 | 2146 | 4125244 |
