siRNA Details
Gene Symbol: EPHA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173641.2 | NP_775912.2 | 3 | 1121 | ||
GenBank mRNA | AJ781169.1 | - | 3 | 1121 | ||
AK090974.1 | - | 3 | 1102 | |||
BC067734.1 | - | 3 | 1121 | 4375803 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
