siRNA Details
Gene Symbol: P4HTM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177938.2 | NP_808807.2 | 3 | 900 | ||
NM_177939.2 | NP_808808.1 | 3 | 900 | |||
GenBank mRNA | AK000269.1 | - | 3 | 304 | ||
AK309082.1 | - | 1 | 480 | |||
AL713728.1 | - | 2 | 1092 | |||
AY198406.1 | - | 3 | 551 | |||
BC000580.1 | - | 1 | 58 | |||
BC011710.2 | - | 3 | 556 | 3940241 | ||
BC047566.1 | - | 3 | 901 | 5259068 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
