siRNA Details
Gene Symbol: HMGB3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002165.1 | 1 | 567 | |||
GenBank mRNA | BC103849.1 | - | 1 | 567 | ||
BC103850.1 | - | 1 | 566 | |||
BC103851.1 | - | 1 | 566 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
