siRNA Details
Gene Symbol: ZNF677 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317998.1 | NP_001304927.1 | 5 | 3501 | ||
NM_182609.3 | NP_872415.1 | 5 | 3319 | |||
XM_005258862.4 | XP_005258919.1 | 5 | 3535 | |||
XM_011526912.2 | XP_011525214.1 | 5 | 3447 | |||
GenBank mRNA | BC050038.1 | - | 5 | 3274 | 5272569 | |
BX648243.1 | - | 5 | 3290 |
