siRNA Details
Gene Symbol: HSD17B7P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF165514.1 | - | 6, 7 | 778 | ||
AF165517.1 | - | 5, 6 | 728 | |||
AK293968.1 | - | 5, 6 | 725 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
