siRNA Details
Gene Symbol: HSD17B7P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF165514.1 | - | 7 | 829 | ||
AF165517.1 | - | 6 | 779 | |||
AK293968.1 | - | 6 | 776 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
