siRNA Details
Gene Symbol: P4HA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288748.1 | NP_001275677.1 | 2 | 375 | ||
NM_182904.4 | NP_878907.1 | 2 | 375 | |||
GenBank mRNA | AK300598.1 | - | 2 | 339 | ||
AL833965.1 | - | 2 | 330 | |||
AY313448.1 | - | 2 | 353 | |||
AY327887.1 | - | 2 | 343 | |||
AY358521.1 | - | 2 | 330 | |||
BC089446.1 | - | 2 | 264 | |||
BC117333.1 | - | 2 | 348 | 40125884 | ||
BC126170.1 | - | 2 | 348 | 8991886 | ||
HY004548.1 | - | 2 | 376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
