siRNA Details
Gene Symbol: PLOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000935.2 | NP_000926.2 | 4, 5 | 1013 | ||
NM_182943.2 | NP_891988.1 | 4, 5 | 1013 | |||
XM_005247535.3 | XP_005247592.1 | 5, 6 | 570 | |||
XM_017006625.1 | XP_016862114.1 | 5, 6 | 524 | |||
GenBank mRNA | AB209879.1 | - | 4, 5 | 685 | ||
AK295084.1 | - | 4, 5 | 604 | |||
BC037169.1 | - | 4, 5 | 684 | 4994235 | ||
U84573.1 | - | 4, 5 | 507 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
