siRNA Details
Gene Symbol: PLOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000935.2 | NP_000926.2 | 13 | 1950 | ||
NM_182943.2 | NP_891988.1 | 13 | 1950 | |||
XM_005247535.3 | XP_005247592.1 | 14 | 1507 | |||
XM_017006625.1 | XP_016862114.1 | 14 | 1461 | |||
GenBank mRNA | AB209879.1 | - | 13 | 1622 | ||
AK125700.1 | - | 3 | 933 | |||
AK295084.1 | - | 13 | 1541 | |||
AL832238.1 | - | 2 | 109 | |||
BC037169.1 | - | 13 | 1621 | 4994235 | ||
BX537521.1 | - | 3 | 1779 | |||
U84573.1 | - | 13 | 1444 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
