siRNA Details
Gene Symbol: ZNF169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301275.1 | NP_001288204.1 | 4 | 368 | ||
NM_003448.2 | NP_003439.2 | 4 | 368 | |||
NM_194320.3 | NP_919301.2 | 4 | 368 | |||
XM_017014362.1 | XP_016869851.1 | 4 | 497 | |||
XM_017014363.1 | XP_016869852.1 | 5 | 391 | |||
XM_017014364.1 | XP_016869853.1 | 5 | 391 | |||
XM_017014365.1 | XP_016869854.1 | 4 | 351 | |||
XM_017014366.1 | XP_016869855.1 | 4 | 309 | |||
GenBank mRNA | AK289742.1 | - | 4 | 312 | ||
BC011947.2 | - | 3 | 288 | |||
BC019228.3 | - | 3 | 288 | |||
BC035060.2 | - | 3 | 288 | |||
BC047702.2 | - | 3 | 292 | 5585128 | ||
BP873364.1 | - | 4 | 323 | |||
DA490480.1 | - | 4 | 336 | |||
HY115642.1 | - | 4 | 368 |
